Order Entry
Czech Republic
ContactUsLinkComponent
Dragendorff's reagent for analysis of alkaloids
Corrosive
Irritant
Dragendorff's reagent for analysis of alkaloids
Catalog # 30989.236
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Irritant
Dragendorff's reagent for analysis of alkaloids
Catalog # 30989.236
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Identification:
    Passes test

Specification Test Results

About this item

 :
Boiling Pt:77 °C (1013 hPa)
Density:1.2 g/cm³ (20 °C)
Flash Pt:–4 °C (closed cup)
Storage Temperature:Ambient
UN:3264
ADR:8,III
 :GHS05
 :GHS07
CAS Number:39775-75-2
MDL Number:MFCD00283106