Order Entry
Czech Republic
ContactUsLinkComponent
Boron trifluoride - methanol complex 20% in methanol for synthesis
Flammable
Toxic
Corrosive
Health
Boron trifluoride - methanol complex 20% in methanol for synthesis
Catalog # 22007.263
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Toxic
Corrosive
Health
Boron trifluoride - methanol complex 20% in methanol for synthesis
Catalog # 22007.263
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Assay:
    13 - 20 %
  • Density (20/4):
    0.880 - 0.895

Specification Test Results

About this item

 :
 :99.85 g/mol
Density:0.89 g/cm³ (20 °C)
Flash Pt:16 °C (closed cup)
EINECS:206-766-4
UN:2924
ADR:3 (8),II
 :GHS02
 :GHS06
 :GHS05
 :GHS08
CAS Number:373-57-9
 :16045-88-8
Formula:CH₄BF₃O
MDL Number:MFCD00071635