Order Entry
Czech Republic
ContactUsLinkComponent
Antimony(III) potassium oxitartrate trihydrate ≥99%, GPR RECTAPUR®
Irritant
Environment
Antimony(III) potassium oxitartrate trihydrate ≥99%, GPR RECTAPUR®
Catalog # 21486.237
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Environment
Antimony(III) potassium oxitartrate trihydrate ≥99%, GPR RECTAPUR®
Catalog # 21486.237
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Cl (Chloride):
    Max. 100 ppm
  • As (Arsenic):
    Max. 0.025 %
  • Fe (Iron):
    Max. 10 ppm

Specification Test Results

About this item

 :
 :667.88 g/mol
Merck Index:13,00706
Melting Pt:100 °C
Density:2.6 g/cm³ (20 °C)
EINECS:229-436-1
UN:1551
ADR:6.1,III
 :GHS07
 :GHS09
CAS Number:28300-74-5
 :304-61-0
Formula:C₈H₄K₂O₁₂Sb₂·3H₂O
MDL Number:MFCD00167056