Order Entry
Czech Republic
ContactUsLinkComponent
Hexafluorosilicic acid 31% in aqueous solution, TECHNICAL
Toxic
Corrosive
Hexafluorosilicic acid 31% in aqueous solution, TECHNICAL
Catalog # 20313.291
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Corrosive
Hexafluorosilicic acid 31% in aqueous solution, TECHNICAL
Catalog # 20313.291
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    Min. 31 %

Specification Test Results

About this item

 :
 :144.09 g/mol
Density:1.29 g/cm³ (20 °C)
EINECS:241-034-8
UN:1778
ADR:8,II
 :GHS06
 :GHS05
CAS Number:16961-83-4
Formula:H₂F₆Si
MDL Number:MFCD00036289