Order Entry
Czech Republic
ContactUsLinkComponent
Silica Analyser Solution No. 1 (metabisulfite/metol)
Irritant
Silica Analyser Solution No. 1 (metabisulfite/metol)
Catalog # 160563U
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Silica Analyser Solution No. 1 (metabisulfite/metol)
Catalog # 160563U
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Silica on reagent:
    Max. 0.01 ppm

Specification Test Results

About this item

 :
 :GHS07