Silica gel 40 - 63 µm for flash chromatography
Catalog # 154425P
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- pH (20°C; 5 %):6.0 - 7.6
- Particle size ( 40 µm):Max. 10 %
- Particle size ( 63 µm):Max. 10 %
Specification Test Results
About this item
:
:60.08 g/mol
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:> 1600 °C
Storage Temperature:Ambient
CAS Number:7631-86-9
:112926-00-8
Formula:SiO₂
MDL Number:MFCD00011232