VWR®, Fuel testing standards - gasoline
Supplier: VWR Chemicals
Certificates
About this item
Fuel testing Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Octane Number.
- Certified in accordance with ASTM / EN or IP test method protocols
- Certified by Round Robin method using independent test laboratories
- Ensures good inter-laboratory correlation
- Supplied in tamper-evident, UN compliant security pack
Frequently Bought Together