VWR®, Silicone Rotational viscosity standards
Supplier: VWR Chemicals
Certificates
About this item
Silicone Rotational viscosity standards are specifically formulated for use with equipment with minimal temperature control. Supplied in 600 ml 'ready to use' packs specially designed to eliminate cleaning of laboratory glassware, which is inherently difficult due to the nature of silicone.
- Ensures full compliance to international test method protocols
- Full traceability to national standards
- Tamper-evident security packaging
- Safer working, less mess, less waste and higher throughput
This range of standards provides both calibration and verification options for rotational viscometer test equipment.
Certifications: Certified according to ISO 17025 / ISO 17034 under UKAS accreditation.
Delivery: 600 ml of product supplied in a ready to use 'test in pack' container.
Caution: Applicable to all spindles with the exception of the L1 spindle, further provision should be made when using the L1 spindle.
The viscosity value given in the ordering table below is the nominal viscosity value - actual batch values are given in the certification documents supplied with the product and also on the label.
The viscosity value given in the ordering table below is the nominal viscosity value - actual batch values are given in the certification documents supplied with the product and also on the label.
Related Information
Certificates
Search by Lot NumberSDS_EN_CH_VWRC99524.pdf SDS_EN_DE_VWRC99524.pdf SDS_EN_NL_VWRC99524.pdf SDS_EN_UK_VWRC99512.pdf SDS_EN_UK_VWRC99513.pdf
SDS
More
VWR® for Calibrations
Product Literature
Frequently Bought Together