VWR®, Cone and Plate viscosity standards
High quality Cone and Plate viscosity standards are ideal for the paints and coatings industry.
- Kinematic viscosity certified in strict accordance with ASTM D2162 at 20, 23 and 25 °C
- Kinematic viscosity certified in accordance with ASTM D341 at intermediate temperatures
- Density, dynamic viscosity and kinematic viscosity quoted at all temperatures
- Ensures full compliance to ASTM and IP test method protocols and had full traceability to national standards
- Tamper-evident security packaging
: Certified according to ISO 17025 / ISO 17034 under UKAS accreditation.
: The viscosity value given in the ordering table below is the nominal viscosity value - actual batch values are given in the certification documents supplied with the product and also on the label.
Frequently Bought Together