ContactUsLinkComponent
USB camera, Moticam® S1
USB camera, Moticam® S1
Catalog # 630-3099
Supplier:  MOTIC
undefined
USB camera, Moticam® S1
Catalog # 630-3099
Supplier:  MOTIC
Specifications
  • Model:
    Moticam S1
  • Resolution:
    1,2 MP
  • Sensor type:
    sCMOS
  • Sensor size:
    1/3"
  • Frame rate:
    1280×960 @ 120 fps; 640×480 @ 240 fps, max.
  • Supported OS:
    Windows 7/8/10, MAC OSX 10.9, Linux or higher
  • Lens mount:
    C-mount
  • Support device:
    TWAIN, SDK and DirectShow Driver
  • Interfaces:
    USB 3.1
  • Sensor:
    sCMOS
  • Software:
    Motic Images Plus 3.0 for Windows, OSX and Linux
  • Pk:
    1
Specifications
About this item
High Grade sCMOS USB camera Moticam® S1 for microscopy for an ultimate imaging result.
  • Fast frame rates
  • sCMOS sensors with back-illuminated sensor technology
  • USB 3.1 interface
  • Motic Images Plus 3.0 software
Moticam S1 is an the entry model for back-illuminated sensor technology and speedy data transfer (with frame rates up to 120 fps*), with easy image setting and capturing. Create your perfect images for a professional database.
Ordering information: To be purchased separately – C-mount adapter not included, this needs to the same brand as your microscope, compatible with your microscope model and fitting the size of your Moticam chip.
Delivery: Includes CS ring adapter, USB 3.1 cable, Motic 4-dot calibration slide, Motic Images Plus 3.0 for Windows/OSX/Linux.
Caution: Minimum computer requirements – 2 GHz dual-core, RAM memory 2 GB, video memory min. 512 MB.