Order Entry
Czech Republic
ContactUsLinkComponent
Tetraethylammonium bromide ≥99%, GPR RECTAPUR®
Tetraethylammonium bromide ≥99%, GPR RECTAPUR®
Catalog # 87084.230
Supplier:  VWR Chemicals
CAS Number:  
undefined
Tetraethylammonium bromide ≥99%, GPR RECTAPUR®
Catalog # 87084.230
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay :
    Min. 99%
  • Water :
    Max. 1,0%

Specification Test Results

About this item

 :
 :210.16 g/mol
Melting Pt:285 °C
 :GHS07
CAS Number:71-91-0
Formula:C₈H₂₀BrN
MDL Number:MFCD00011825