ESD safety ankle boots, lace-up, L10, Vintage Pirate
: ELTEN
Vintage style ESD safety shoe made from waxy cowhide leather, with padded tongue and a dual-density VIBRAM® rubber/PU tread sole L10 PIRATE.
- Fabric lining Bioactive
- Replaceable insole; orthopaedic insoles possible
- Full-length inlay sole ESD PRO grey
- Anatomically shaped insole
- Metal-free, puncture resistant midsole
- Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Grey/red
: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC
- Catalog No:
- 114-8838
- 114-8839
- 114-8836
- 114-8837
- 114-8832
- 114-8840
- 114-8842
- 114-8841
- 114-8834
- 114-8835
- 114-8833
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
- ISO 14001
Frequently Bought Together