Human Recombinant GDF-15 (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Growth differentiation factor 15 (GDF-15) is a member of the transforming growth factor beta (TGF-β) family and is made by the placenta and cardiovascular tissues. GDF-15 regulates inflammatory and apoptotic pathways during cellular stress and injury. GDF-15 is emerging as a biomarker of early heart disease, such that increased levels of circulating GDF-15 are associated with an increased risk of developing heart failure.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q99988
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:MIC-1|Prostate differentiation factor|Placental TGF-β
- Protein/peptide name:GDF-15 (AF)
- Purity:≥95%
- Molecular weight:12,4/24,8 kDa
- Sequence:MARNGDHCPL GPGRCCRLHT VRASLEDLGW ADWVLSPREV QVTMCIGACP SQFRAANMHA QIKTSLHRLK PDTVPAPCCV PASYNPMVLIQKTDTGVSLQ TYDDLLAKDC HCI
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together