Mouse Recombinant MIF (from E. coli)
Macrophage migration inhibitory factor (MIF) is a pro-inflammatory lymphokine that functions during cell-mediated immmunity. MIF promotes fibroblast migration by inducing interleukin 1 (IL-1), interleukin 8 (IL-8), and matrix metalloproteinase (MMP) expression. In interferon-gamma-activated macrophages, MIF stimulates nitric oxide (NO) production and tumor necrosis factor alpha (TNF-α) secretion.
- High quality
- Low endotoxin
- Affordable
: This product is for research use only and is not for use in humans.
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P34884
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein/peptide name:MIF (AF)
- Purity:≥95%
- Molecular weight:12,5 kDa
- Sequence:MPMFIVNTNV PRASVPEGFL SELTQQLAQA TGKPAQYIAV HVVPDQLMTF SGTNDPCALC SLHSIGKIGG AQNRNYSKLL CGLLSDRLHI SPDRVYINYY DMNAANVGWN GSTFA
- Endotoxin level:Low
- Formulation:10 mM Sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Frequently Bought Together