Mouse Recombinant IL-17E (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Interleukin 17E (IL-17E), also known as IL-25, is a member of the IL-17 family of cytokines. IL-17E binds to the IL-17RB receptor to stimulate the secretion of the proinflammatory interleukin 8 (IL-8) chemokine and to induce the activation of nuclear factor kappa-light-chain-enhancer of activated B cells (NF-κB).
- High quality
- Low endotoxin
- Affordable
Caution: This product is for research use only and is not for use in humans.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q8VHC9
- Reconstitution instructions:Sterile 10 mM HCl at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:IL-25
- Protein/peptide name:IL-17E (AF)
- Purity:≥90%
- Molecular weight:17,6/35,2 kDa
- Sequence:MVSLRIQEGC SHLPSCCPSK EQEPPEEWLK WSSASVSPPE PLSHTHHAES CRASKDGPLN SRAISPWSYE LDRDLNRVPQ DLYHARCLCP HCVSLQTGSH MDPLGNSVPL YHNQTVFYRR PCHGEEGTHR RYCLERRLYR VSLACVCVRP RVMA
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together