Order Entry
Czech Republic
ContactUsLinkComponent
Human Recombinant Omentin (Animal Free) (from E. coli)
Human Recombinant Omentin (Animal Free) (from E. coli)
Certificates
undefined
Human Recombinant Omentin (Animal Free) (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤5
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    Q8WWA0
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    Intelectin-1|ITLN-1
  • Protein/peptide name:
    Omentin
  • Purity:
    ≥90%
  • Molecular weight:
    35 kDa
  • Sequence:
    MNQLSFLLFL IATTRGWSTD EANTYFKEWT CSSSPSLPRS CKEIKDECPS AFDGLYFLRT ENGVIYQTFC DMTSGGGGWT LVASVHENDM RGKCTVGDRW SSQQGSKADY PEGDGNWANY NTFGSAEAAT SDDYKNPGYY DIQAKDLGIW HVPNKSPMQH WRNSSLLRYR TDTGFLQTLG HNLFGIYQKY PVKYGEGKCW TDNGPVIPVV YDFGDAQKTA SYYSPYGQRE FTAGFVQFRV FNNERAANAL CAGMRVTGCN TEHHCIGGGG YFPEASPQQC GDFSGFDWSG YGTHVGYSSS REITEAAVLL FYR
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate and 5:1 mannitol to protein, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Related Information

Frequently Bought Together