Human Recombinant EBI3 (Animal free) (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Epstein-Barr virus induced gene-3 (EBI3) is a secreted glycoprotein belonging to the hematopoietin receptor family related to the p40 subunit of interleukin 12 (IL-12). EBI3 expression is induced in B-lymphocytes in response to Epstein-Barr virus infection. EBI3 forms heterodimers with p28 to form interleukin 27 (IL-27), and with p35 to form interleukin 35 (IL-35). Both IL-27 and IL-35 have anti-inflammatory and regulatory activity.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Delivery: A certificate of analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤5
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q14213
- Reconstitution instructions:Sterile 10 mM HCl at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:IL-35/EBI3|IL-27/EBI3
- UniProtKB:Q14213
- Protein/peptide name:EBI3 subunit (IL-27/IL-35)
- Purity:≥90%
- Molecular weight:23,4 kDa
- Sequence:MRKGPPAALT LPRVQCRASR YPIAVDCSWT LPPAPNSTSP VSFIATYRLG MAARGHSWPC LQQTPTSTSC TITDVQLFSM APYVLNVTAV HPWGSSSSFV PFITEHIIKP DPPEGVRLSP LAERQLQVQW EPPGSWPFPE IFSLKYWIRY KRQGAARFHR VGPIEATSFI LRAVRPRARY YVQVAAQDLT DYGELSDWSL PATATMSLGK
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic Acid (TFA) + 0,5% mannitol
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together