Human Recombinant RANK LIGAND (Animal free) (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Stromal cell-derived factor-1 alpha (SDF-1 α), also called CXCL12a, is one of two SDF-1 splice variants made by a wide variety of cells upon stimulation by inflammatory cytokines such as TNF, IL-1, and LPS. SDF-1α signals through the G protein-coupled receptor CXCR4 to recruit activated leukocytes. Human and mouse SDF-1 α share 99% sequence identity.
- High quality
- Low endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Delivery: Certificate of Analysis (COA) is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity assay: Ability to induce bioluminescence in engineered RANKL bioassay cells , ED50 ≤50 ng/ml, bioactivity ≥2,0×10⁴ units/mg
- Gene ID:O14788
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:OPGL|TNFSF11|ODF|TRANCE
- UniProtKB:O14788
- Protein/peptide name:RANK Ligand
- Purity:≥95%
- Molecular weight:19,7 kDa
- Sequence:EKAMVDGSWL DLAKRSKLEA QPFAHLTINA TDIPSGSHKV SLSSWYHDRG WAKISNMTFS NGKLIVNQDG FYYLYANICF RHHETSGDLA TEYLQLMVYV TKTSIKIPSS HTLMKGGSTK YWSGNSEFHF YSINVGGFFK LRSGEEISIE VSNPSLLDPD QDATYFGAFK VRDID
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together