


Methanol, anhydrous 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
:32.04 g/mol
Merck Index:14,5957
Boiling Pt:64.7 °C
Melting Pt:–98 °C
Density:0.791
EINECS:200-659-6
UN:1230
ADR:3,II
:Packaged under argon.
Flash Pt:11 °C (52 °F)
:GHS02
:GHS06
:GHS08
Storage Temperature:Ambient
Frequently Bought Together