


Zinc chloride, anhydrous ≥98%, Reagent Grade, free-flowing, Fluka™
Catalog # HONC793523-25KG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 kg
- Seq:4283
Specifications
Specification Test Results
- Assay:Min. 97.0 %
- Insoluble in HCl:Max. 0.005 %
- Calcium (Ca):Max. 0.06 %
- Iron (Fe):Max. 0.001 %
- Potassium (K):Max. 0.02 %
- Magnesium (Mg):Max. 0.01 %
- Sodium (Na):Max. 0.05 %
- Lead (Pb):Max. 0.005 %
- Oxide chloride (as ZnO):complying
- Ammonium (NH₄):Max. 0.005 %
- Nitrate (NO₃):Max. 0.003 %
- Sulfate (SO₄):Max. 0.01 %
- Appearance (color):complying
- Appearance (form):complying
Specification Test Results
About this item
:
:136.3 g/mol
Density:2.93 g/cm3
UN:2331
ADR:8,III
:GHS09
:GHS07
:GHS05
CAS Number:7646-85-7
Formula:ZnCl₂
MDL Number:MFCD00011295