Order Entry
Czech Republic
ContactUsLinkComponent
Tetrahydrofuran, anhydrous ≥99.9% unstabilised for applications requiring low water content <0.0025% H₂O, Riedel-de Haën™
Flammable
Health
Irritant
Tetrahydrofuran, anhydrous ≥99.9% unstabilised for applications requiring low water content <0.0025% H₂O, Riedel-de Haën™
Catalog # HONC66205-2.5L
CAS Number:  
undefined
Flammable
Health
Irritant
Tetrahydrofuran, anhydrous ≥99.9% unstabilised for applications requiring low water content <0.0025% H₂O, Riedel-de Haën™
Catalog # HONC66205-2.5L
CAS Number:  

Specifications

  • Pack type:
    Bottle, septum-sealed
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.9 %
  • Water (Karl Fischer):
    Max. 0.0025 %
  • Non-volatile matter:
    Max. 0.0001 %

Specification Test Results

About this item

 :
 :72.11 g/mol
Boiling Pt:65 °C (1.013 hPa)
Melting Pt:–108 °C
Density:0.89 g/cm3 (20 °C)
Flash Pt:–21 °C
Storage Temperature:Ambient
EINECS:203-726-8
UN:2056
ADR:3,II
 :Packed in septum-sealed bottle.
 :GHS02
 :GHS08
 :GHS07
CAS Number:109-99-9
Formula:C₄H₈O
MDL Number:MFCD00005356