


N,N-Dimethylformamid, anhydrous ≥99.9% for applications requiring low water content, over molecular sieves <0.0050% H₂O, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:68-12-2
:
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284
Boiling Pt:152…153 °C (1,013 hPa)
Melting Pt:–61 °C
Density:0,94 g/cm3 (20 °C)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
:Packed in septum-sealed bottle.
:GHS07
:GHS08
:GHS02
Specification Test Results
- Assay (GC):Min. 99.9 %
- Water (Karl Fischer):Max. 0.0050 %
Specification Test Results
Frequently Bought Together