ContactUsLinkComponent
Tetrahydrofuran, anhydrous ≥99.9% unstabilised for applications requiring low water content, over molecular sieves <0.0025% H₂O, Riedel-de Haën™
Flammable
Health
Irritant
Tetrahydrofuran, anhydrous ≥99.9% unstabilised for applications requiring low water content, over molecular sieves <0.0025% H₂O, Riedel-de Haën™
Certificates
undefined
Flammable
Health
Irritant
Tetrahydrofuran, anhydrous ≥99.9% unstabilised for applications requiring low water content, over molecular sieves <0.0025% H₂O, Riedel-de Haën™
Specification Test Results
  • Assay (GC):
    Min. 99.9 %
  • Water (Karl Fischer):
    Max. 0.0025 %