Order Entry
Czech Republic
ContactUsLinkComponent
D-(+)-Sucrose, Extra pure meets analytical specifications of Reagent Ph. Eur., BP, NF, Burdick & Jackson™
D-(+)-Sucrose, Extra pure meets analytical specifications of Reagent Ph. Eur., BP, NF, Burdick & Jackson™
Catalog # HONC65893-10KG
CAS Number:  
undefined
D-(+)-Sucrose, Extra pure meets analytical specifications of Reagent Ph. Eur., BP, NF, Burdick & Jackson™
Catalog # HONC65893-10KG
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 kg
  • Seq:
    4158

Specifications

Specification Test Results

  • Identity:
    complying
  • Specific rotation (a)20/D;C=26 in H₂O:
    66.3 - 67.0 °
  • Conductivity 20 °C:
    Max. 35 µS/cm
  • Loss on drying (105°C, 3h):
    Max. 0.1 %
  • Residue on ignition (SO₄):
    Max. 0.05 %
  • Acidity or alkalinity:
    complying
  • Calcium (Ca):
    complying
  • Lead (Pb):
    Max. 0.5 ppm
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 0.0035 %
  • Sulfate (SO₄):
    Max. 0.006 %
  • Sulfite (as SO₂):
    Max. 0.001 %
  • Dextrines:
    complying
  • Invert sugar:
    complying
  • Reducing sugars:
    complying
  • Colour index:
    Max. 45
  • Appearance of the solution:
    complying
  • Residual solvents:
    complying
  • Endotoxin limit (LAL):
    Max. 250 EU/g

Specification Test Results

About this item

 :
Melting Pt:185-186 °C
Melting Pt:185…186 °C
Density:1.580 g/cm3 (20 °C)
 :342.3 g/mol
Storage Temperature:Ambient
EINECS:200-334-9
CAS Number:57-50-1
Formula:C₁₂H₂₂O₁₁
MDL Number:MFCD00006626