Order Entry
Czech Republic
ContactUsLinkComponent
n-Hexane, for GC Plus, Burdick & Jackson™
n-Hexane, for GC Plus, Burdick & Jackson™
Catalog # HONC65829-7L-RC
CAS Number:  
undefined
n-Hexane, for GC Plus, Burdick & Jackson™
Catalog # HONC65829-7L-RC
CAS Number:  

Specifications

  • Pack type:
    Stainless steel can
  • Pk:
    7 L
  • Seq:
    5202

Specifications

Specification Test Results

  • Assay (GC):
    Min. 95 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Suitability for residue analysis:
    Max. 5 ng/l
  • Suitability for GC-MS of dioxines, furanes, PCPs:
    complying

Specification Test Results

About this item

 :
 :86.18 g/mol
Boiling Pt:66…69 °C (1.013 hPa)
Melting Pt:–95 °C
Density:0.660…0.668 g/cm3 (20 °C)
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
 :GHS09
 :GHS07
 :GHS08
 :GHS02
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520