Order Entry
Czech Republic
ContactUsLinkComponent
 
Environment
Irritant
Health
Flammable
Iso-Hexane ≥85% (by GC, mixture of isomers) for gas chromatography, Burdick & Jackson™
Catalog # HONC65824-200L-RC
CAS Number:  
Environment
Irritant
Health
Flammable
Iso-Hexane ≥85% (by GC, mixture of isomers) for gas chromatography, Burdick & Jackson™
Catalog # HONC65824-200L-RC
CAS Number:  

Specifications

  • Pack type:
    Stainless steel drum, returnable
  • Pk:
    200 L
  • Seq:
    5549

Specifications

Specification Test Results

  • Assay of isomers (GC):
    Min. 85 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Suitability for residue analysis:
    Max. 5 ng/l
  • n-Hexane (GC):
    Max. 4.9 %

Specification Test Results

About this item

 :
 :86.18 g/mol
Boiling Pt:48…65 °C
Density:0.66 g/cm3
Flash Pt:–26 °C
UN:1208
ADR:3,II
 :GHS09
 :GHS07
 :GHS08
 :GHS02
CAS Number:64742-49-0
Formula:C₆H₁₄