Order Entry
Czech Republic
ContactUsLinkComponent
n-Hexane ACS, Reag. Ph. Eur., Burdick & Jackson™
n-Hexane ACS, Reag. Ph. Eur., Burdick & Jackson™
Catalog # HONC65801-200L
CAS Number:  
n-Hexane ACS, Reag. Ph. Eur., Burdick & Jackson™
Catalog # HONC65801-200L
CAS Number:  

Specifications

  • Pack type:
    Metal drum
  • Pk:
    200 L
  • Seq:
    5549

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99 %
  • Boiling range:
    68 - 69 °C
  • Boiling range 67-69 °C:
    Min. 95 %
  • Density (D 20/4):
    0.658 - 0.662
  • Density (D 20/20):
    0.659 - 0.663
  • Refractive index (n 20/D):
    1.3745 - 1.3755
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Free acid (as CH₃COOH):
    Max. 0.002 %
  • Aluminium (Al):
    Max. 0.00005 %
  • Boron (B):
    Max. 0.000002 %
  • Barium (Ba):
    Max. 0.00001 %
  • Calcium (Ca):
    Max. 0.00005 %
  • Cadmium (Cd):
    Max. 0.000005 %
  • Cobalt (Co):
    Max. 0.000002 %
  • Chromium (Cr):
    Max. 0.000002 %
  • Copper (Cu):
    Max. 0.000002 %
  • Iron (Fe):
    Max. 0.00001 %
  • Magnesium (Mg):
    Max. 0.00001 %
  • Manganese (Mn):
    Max. 0.000002 %
  • Nickel (Ni):
    Max. 0.000002 %
  • Lead (Pb):
    Max. 0.00001 %
  • Tin (Sn):
    Max. 0.00001 %
  • Zinc (Zn):
    Max. 0.00001 %
  • Aromatic substances (as C₆H₆):
    Max. 0.01 %
  • S-compounds (as S):
    Max. 0.005 %
  • Thiophene:
    Max. 0.0001 %
  • Reaction against H₂SO₄:
    complying
  • APHA:
    Max. 10

Specification Test Results

About this item

 :
 :86.18 g/mol
Boiling Pt:66…69 °C
Density:0.660…0.668 g/cm3
Flash Pt:–26 °C
UN:1208
ADR:3,II
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520