


Tetrahydrofuran stabilised ACS, Burdick & Jackson™
Catalog # HONC65777-200L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Metal drum
- Pk:200 L
- Seq:5549
Specifications
Specification Test Results
- Assay (GC):Min. 99.9 %
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.05 %
- Free acid (as CH₃COOH):Max. 0.001 %
- Aluminium (Al):Max. 0.00005 %
- Boron (B):Max. 0.000002 %
- Barium (Ba):Max. 0.00001 %
- Calcium (Ca):Max. 0.00005 %
- Cadmium (Cd):Max. 0.000005 %
- Cobalt (Co):Max. 0.000002 %
- Chromium (Cr):Max. 0.000002 %
- Copper (Cu):Max. 0.000002 %
- Iron (Fe):Max. 0.00001 %
- Magnesium (Mg):Max. 0.00001 %
- Manganese (Mn):Max. 0.000002 %
- Nickel (Ni):Max. 0.000002 %
- Lead (Pb):Max. 0.00001 %
- Tin (Sn):Max. 0.00001 %
- Zinc (Zn):Max. 0.00001 %
- Peroxides (as H₂O₂):Max. 0.001 %
- Assay BHT (GC):100 - 300 ppm
- APHA:Max. 20
Specification Test Results
About this item
:
:72.11 g/mol
Boiling Pt:65 °C
Density:0.89 g/cm3
Flash Pt:–21 °C
UN:2056
ADR:3,II
:GHS08
:GHS07
:GHS02
CAS Number:109-99-9
Formula:C₄H₈O
MDL Number:MFCD00005356