Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
EDTA disodium salt dihydrate 99.0-101.0%, meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™
 
undefined
 
 
EDTA disodium salt dihydrate 99.0-101.0%, meets analytical specification of BP, Ph. Eur., USP, FCC, Fluka™

 

  • Identity (IR):
    complying
  • Assay (calculated to the dried substance):
    99.0 - 101.0 %
  • Loss on drying (150°C):
    8.7 - 11.4 %
  • pH (1 %,25°C):
    4.3 - 4.7
  • pH (5 %, 25 °C):
    4.0 - 6.0
  • pH (5 %, 20°C):
    4.0 - 5.0
  • Arsenic (As):
    Max. 0.0001 %
  • Calcium (Ca):
    complying
  • Iron (Fe):
    Max. 10 ppm
  • Lead (Pb):
    Max. 10 ppm
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Chloride (Cl):
    Max. 0.01 %
  • Cyanide (CN):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.05 %
  • Nitrilotriacetic acid (HPLC):
    Max. 0.1 %
  • Residual solvents:
    complying
  • Appearance of the solution:
    complying

 

Frequently Bought Together