ContactUsLinkComponent
Delivery tubes for OTAL® disposable pumps
Delivery tubes for OTAL® disposable pumps
Catalog # 300-0361
Supplier:  BURKLE
Delivery tubes for OTAL® disposable pumps
Catalog # 300-0361
Supplier:  BURKLE
Specifications
  • Description:
    Delivery tubes
  • Material:
    PE
  • Immersion depth:
    90 cm
  • Pk:
    10
  • Tube Ø:
    12 mm
Specifications
About this item
Disposable delivery tube made of PE in a pack of 10, can be cut to length.