Tetrahydrofuran, anhydrous ≥99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Butylene oxide, THF, Diethylene oxide
CAS Number:109-99-9
:
Formula:C₄H₈O
MDL Number:MFCD00005356
Boiling Pt:66 °C
Melting Pt:–108.4°C
Density:0.889
Molecular Weight:72.11 g/mol
Flash Pt:–21°C
EINECS:203-726-8
UN:2056
ADR:3 II
Merck Index:12,09351
:Packaged under argon.
:GHS02
:GHS07
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless liquid
- Assay (GC):≥99.8%
- Water content (Karl Fischer Titration):≤50 ppm
- Stabilizer:BHT
Specification Test Results
Frequently Bought Together