General purpose gloves, nitrile, Showa S-TEX 376R
Dodavatel: Showa
Certifikáty
O tomto produktu
Showa S-TEX 376 durable double-dipped nitrile coated glove that provides resistance to oil whilst protecting from cuts and lacerations. Ideal for the secure handling of oily components and materials that present a high degree of mechanical risk. Manufactured with SHOWA's patented Hagane Coil® technology.
- ¾ coated nitrile with an additional coating of foam nitrile on the palm, fingertips and knuckles
- Protects the hand from oils, hydrocarbons, grease and abrasion
- Long lasting grip performance in wet and oily conditions
- Anatomical hand shape design helps prevent hand fatigue
- Seamless nylon/polyester knitted liner
- High mechanical resistance to abrasion
Idea for use in aerospace, automotive, construction, oil and gas, utilities and waste management. Typical application includes assembly, handling oily/greasy components, maintenance, metal fabrication, mixing mortar and refuse collection.
Certifications: EN 388:2016 (4121X); Cat. II
Specifikace
- Catalog No:
- 112-6011
- 112-6007
- 112-6008
- 112-6009
- 112-6010
- Model:
- S-TEX 376
- S-TEX 376
- S-TEX 376
- S-TEX 376
- S-TEX 376
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Nylon/polyester
- Nylon/polyester
- Nylon/polyester
- Nylon/polyester
- Nylon/polyester
- Finish:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
Specifikace
Frequently Bought Together