Order Entry
Czech Republic
ContactUsLinkComponent
Orthophosphoric acid 49.5 - 50.5%, LiChropur™ for HPLC, Supelco®
Corrosive
Orthophosphoric acid 49.5 - 50.5%, LiChropur™ for HPLC, Supelco®
Catalog # 93752-250ML
Supplier:  Merck
CAS Number:  
Corrosive
Orthophosphoric acid 49.5 - 50.5%, LiChropur™ for HPLC, Supelco®
Catalog # 93752-250ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Appearance (Color):
    Colorless
  • Appearance (Form):
    Liquid
  • Titration (T) NaOH 1M:
    49.5 - 50.5 %
  • Suitability Test (HPLC):
    Corresponds to Standard
  • Density D20/4:
    1.331 - 1.336
  • UV-Absorption:
    230nm ≤ 0.03, 230nm ≤ 0.03
  • Test by UV Spectroscopy:
    240nm ≤ 0.02, 280nm ≤ 0.02, 500nm ≤ 0.02
  • Residue (Filter Test):
    No Residue
  • Filtertest (Method):
    Neat
  • Recommended Retest Period:
    54 Months

Specification Test Results

About this item

 :
 :98 g/mol
Density:1.331...1.336 g/cm³ (20 °C)
UN:1805
ADR:8,III
 :GHS05
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340