



Boron trichloride solution 11,0 - 13,0% (by alkalimetry) in methanol, LiChropur™ for GC derivatisation, Supelco®
: Merck
:10294-34-5
:
:BCl₃
:MFCD00011313
:117.17 g/mol
:0.875...0.895 g/m³ (20 °C)
:3286
:3 (6.1, 8),II
:GHS02
:GHS05
:GHS06
:GHS08
- Appearance (Color):Colorless
- Appearance (Form):Liquid
- Titration:11.0 - 13.0 %
- Titration (Method):Alkalimetric
- Remarks on GC:Suitibility for GC derivatization complying
- Density D20/4:0.875 - 0.895
- Infrared Spectrum:Conforms to Structure
- Recommended Retest Period:5 Months
Frequently Bought Together