Order Entry
Czech Republic
ContactUsLinkComponent
Ammonium sulphamate ≥98.5%, AnalaR NORMAPUR® ACS for analysis, for detection of sulphonamide in blood
Ammonium sulphamate ≥98.5%, AnalaR NORMAPUR® ACS for analysis, for detection of sulphonamide in blood
Catalog # 85979.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ammonium sulphamate ≥98.5%, AnalaR NORMAPUR® ACS for analysis, for detection of sulphonamide in blood
Catalog # 85979.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay:
    Min. 98,5 %
  • Melting point:
    131,0 - 135,0 °C
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Insoluble matter:
    Max. 0,02 %
  • Residue after ignition:
    Max. 0,10 %

Specification Test Results

About this item

 :
 :114.13 g/mol
CAS Number:7773-06-0
Formula:H₆N₂O₃S
MDL Number:MFCD00011429