Order Entry
Czech Republic
ContactUsLinkComponent
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) ≥99.0% (by GC) for ion chromatography, Supelco®
Irritant
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) ≥99.0% (by GC) for ion chromatography, Supelco®
Catalog # 80833-10G
Supplier:  Merck
CAS Number:  
Irritant
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) ≥99.0% (by GC) for ion chromatography, Supelco®
Catalog # 80833-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance (Color):
    White to Faint Yellow
  • Appearance (form):
    Powder or Solid or Crystals or Crystalline Chunk(s) or Chunk(s)
  • Purity (GC area %):
    ≥ 99.0 %
  • Carbon Content:
    54.0 - 55.1 %
  • Infrared Spectrum:
    Conforms to Structure
  • Metal Trace Analysis (ICP):
    Corresponds to Requirements
  • Aluminium (ICP):
    ≤ 1 mg/kg
  • Barium (ICP):
    ≤ 1 mg/kg
  • Bismuth (ICP):
    ≤ 1 mg/kg
  • Calcium (ICP):
    ≤ 5 mg/kg
  • Cadmium (ICP):
    ≤ 1 mg/kg
  • Cobalt (ICP):
    ≤ 1 mg/kg
  • Chromium (ICP):
    ≤ 1 mg/kg
  • Copper (ICP):
    ≤ 1 mg/kg
  • Iron (ICP):
    ≤ 1 mg/kg
  • Potassium (ICP):
    ≤ 20 mg/kg
  • Lithium (ICP):
    ≤ 1 mg/kg
  • Magnesium (ICP):
    ≤ 1 mg/kg
  • Manganese (ICP):
    ≤ 1 mg/kg
  • Molybdenum (ICP):
    ≤ 1 mg/kg
  • Sodium (ICP):
    ≤ 20 mg/kg
  • Nickel (ICP):
    ≤ 1 mg/kg
  • Lead (ICP):
    ≤ 1 mg/kg
  • Strontium (ICP):
    ≤ 1 mg/kg
  • Zinc (ICP):
    ≤ 1 mg/kg
  • Ion Chromatography (NH₄⁺):
    ≤ 10 mg/kg

Specification Test Results

About this item

 :
 :264.32 g/mol
Melting Pt:42...45 °C (lit.)
EINECS:241-473-5
 :GHS07
CAS Number:17455-13-9
Formula:C₁₂H₂₄O₆
MDL Number:MFCD00005113