EDTA disodium salt solution with zinc complex added (solution A) volumetric solution, for hardness of water determinations (1 ml = 5.6°dH/100 ml H₂O), Fluka™
Catalog # 34547-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Factor:0.999 - 1.001
Specification Test Results
About this item
:
Density:1.0 g/cm3
Storage Temperature:Ambient