
N-(1-Naphthyl)ethylenediamine dihydrochloride ≥98% ACS for determination of sulphonamide and nitrite, Fluka™
Catalog # HONC33461H-5G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Water (Karl Fischer):max. 5 %
- Suitability for detect. of Sulfonamide:complying
- Solubility in water:complying
- 1-Naphthylamine:max. 0.1 %
- 2-Naphthylamine:max. 0.01 %
- Identity:complying
Specification Test Results
About this item
:
:259.18 g/mol
Merck Index:12,06496
Melting Pt:194-198 °C (dec.)
EINECS:215-981-2
:GHS07
Storage Temperature:Ambient
CAS Number:1465-25-4
Formula:C₁₂H₁₆Cl₂N₂
MDL Number:MFCD00012556
Melting Pt:194…198 °C (dec.)