Order Entry
Czech Republic
ContactUsLinkComponent
Sodium tetrakis(4-fluorophenyl)borate dihydrate ≥97.0%, Selectophore™ ionophore, Supelco®
Irritant
Sodium tetrakis(4-fluorophenyl)borate dihydrate ≥97.0%, Selectophore™ ionophore, Supelco®
Catalog # 72014-500MG
Supplier:  Merck
CAS Number:  
undefined
Irritant
Sodium tetrakis(4-fluorophenyl)borate dihydrate ≥97.0%, Selectophore™ ionophore, Supelco®
Catalog # 72014-500MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :450.215 g/mol
 :M-Clarity™ quality level = MQ100
 :GHS07
CAS Number:207683-22-5
Formula:C₂₄H₂₀BF₄NaO₂
MDL Number:MFCD00149598