Order Entry
Czech Republic
ContactUsLinkComponent
Nucleic acid isolation kits, SeraSil-Mag™ beads
Nucleic acid isolation kits, SeraSil-Mag™ beads
Supplier:  Cytiva
Certificates
undefined
Nucleic acid isolation kits, SeraSil-Mag™ beads
Supplier:  Cytiva

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Blood
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    SeraSil-Mag™ 700

Specifications

Frequently Bought Together