

1,2-Dichlorobenzene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:95-50-1
:
Formula:C₆H₄Cl₂
MDL Number:MFCD00000535
:147 g/mol
UN:1591
ADR:6.1,III
:GHS07
:GHS09
Specification Test Results
- Assay:Min. 99%
- Density (20/4):1.305 - 1.307
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC85995.180 spec_VWRC85995.290 spec_VWRC85995.320
Product specification
Frequently Bought Together