Order Entry
Czech Republic
ContactUsLinkComponent
 
Peqlab peqGOLD, DNA Isolation, Microspin Tissue DNA Mini Kits
Catalog # 13-3096-02
Supplier:  VWR Chemicals
CAS Number:  
undefined
Peqlab peqGOLD, DNA Isolation, Microspin Tissue DNA Mini Kits
Catalog # 13-3096-02
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    200 Tests
  • Application:
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Micro
  • Refine target molecule:
    Total DNA
  • Sample type:
    Swabs
    Forensic samples
    Blood (dried) on filter cards
    Blood
    Biofluids (serum, plasma, saliva)
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    Microspin tissue DNA mini kit
  • Seq:
    03
  • Environmentally Preferable:
  • Sample size:
    10 mg of tissue or 1 - 100 ml of fluid
  • Description:
    Microspin tissue DNA mini kit

Specifications

About this item

Fast, easy, and economical kit for isolation of genomic DNA from small sample volumes and sizes for consistent PCR and analytical test results.

  • No need for organic extractions
  • No need for time-consuming ethanol precipitations
  • Enzymatic degradation of cellular RNA
  • Efficient separation of inhibitors and contaminants
  • Fast, 20 min or less

Typical starting amount 10 mg tissue. Speed of mini column technology and specifically formulated buffers allow the purification of genomic DNA up to 60 kb.

Delivery: Kit contains peqGOLD Microspin LE DNA columns, 2 ml collection tubes, BL buffer, TL buffer, Proteinase K solution, HBC buffer, LPA buffer DNA wash buffer and elution buffer.