Order Entry
Czech Republic
ContactUsLinkComponent
Dodecyltrimethylammonium bromide ≥99%, GPR RECTAPUR®
Dodecyltrimethylammonium bromide ≥99%, GPR RECTAPUR®
Catalog # 85982.130
Supplier:  VWR Chemicals
CAS Number:  
undefined
Dodecyltrimethylammonium bromide ≥99%, GPR RECTAPUR®
Catalog # 85982.130
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Assay :
    Min. 99 %
  • IR Spectrum:
    Conforms to structure
  • Colouration (10 %; water):
    Max. 100 APHA
  • Solubility in water (10 %):
    Clear to slightly hazy

Specification Test Results

About this item

 :
 :308.35 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,II
 :GHS06
 :GHS09
CAS Number:1119-94-4
Formula:C₁₅H₃₄BrN
MDL Number:MFCD00011767