



2-Pyridylzinc bromide solution 0.5 M in THF
Catalog # H58544.AD
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:50 mL
- Seq:2370
Specifications
About this item
:
:223.378 g/mol
Density:0.974
ADR:4.3,II
:Packaged under argon in resealable ChemSeal™ bottles.
Storage Temperature:Refrigerator
UN:3399
:GHS02
:GHS05
:GHS08
:GHS07
CAS Number:218777-23-2
Formula:C₅H₄BrNZn
MDL Number:MFCD00671985