Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lewatit® AF 5
Lewatit® AF 5, Pore size: 20 Å (mean), 20-50 mesh, Supelco®
Lewatit® AF 5, Pore size: 20 Å (mean), 20-50 mesh, Supelco®
Catalog # 52089-500G-F
Supplier:
Merck
CAS Number:
Expand image
Lewatit® AF 5, Pore size: 20 Å (mean), 20-50 mesh, Supelco®
Catalog # 52089-500G-F
Supplier:
Merck
CAS Number:
Specifications
Pk:
500 g
Seq:
3645
Specifications
About this item
Product Details
:
Product Details