ContactUsLinkComponent
TISAB II Solution, Supelco®
TISAB II Solution, Supelco®
Katalogové číslo 89466-500ML
Supplier:  Merck
CAS Number:  
TISAB II Solution, Supelco®
Katalogové číslo 89466-500ML
Supplier:  Merck
CAS Number:  
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    TISAB II solution
  • M-Clarity quality level:
    MQ100
Specifications
About this item
TISAB, in general, is mixed with samples, as it helps in maintaining the ionic strength and pH constant. It is mainly used to decomplex metal-F complexes.
TISAB II can be prepared using NaCl with glacial acetic acid, 1,2-Cyclohexylenedinitrilotetraacetic acid and NaOH. It also has high concentrations of cation chelators that hydrolyse fluoride-ion pairs to form fluoride ions.
TISAB II has been used to determine total fluoride concentration using fluoride ion sensitive electrode from dental varnish.