ContactUsLinkComponent
TISAB I Solution, Supelco®
TISAB I Solution, Supelco®
Catalog # 89465-500ML
Supplier:  Merck
CAS Number:  
TISAB I Solution, Supelco®
Catalog # 89465-500ML
Supplier:  Merck
CAS Number:  
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    TISAB I solution
  • M-Clarity quality level:
    MQ100
Specifications
About this item
TISAB, in general, is mixed with samples, as it helps in maintaining the ionic strength and pH constant. It is mainly used to decomplex metal-F complexes.
TISAB I can be prepared using NaCl with glacial acetic acid, tri-sodium citrate and NaOH.
TISAB I has been used in determination of fluoride-ion concentration using fluoride ion sensitive electrode.