ContactUsLinkComponent
MALDI validation set polystyrene Mp ~700 - 70000, certified reference material, Supelco®
MALDI validation set polystyrene Mp ~700 - 70000, certified reference material, Supelco®
  03565-1EA
 :  Merck
 :  
MALDI validation set polystyrene Mp ~700 - 70000, certified reference material, Supelco®
  03565-1EA
 :  Merck
 :  
  • Pk:
    1
  • Standards:
    Certified reference material
  • CAS No.:
    9003-53-6
  • Description:
    MALDI validation set polystyrene Mp ~700 - 70 000
  • M-Clarity quality level:
    MQ100
  • Description:
    MALDI validation set polystyrene Mp ~700 - 70 000
MALDI Standards are suitable for the validation and comparison of MALDI-ToF instruments. Certificates provide MALDI spectra including Mw, Mn and Mp values of select narrow polymers, which differ in molar mass.
This set inclces six different polystyrene standards ranging from Mp ~700 to ~70000.
Each standard is characterized with GPC, a second absolute method (light scattering or H-NMR) and MALDI-ToF-MS.