Sodium cyanide ≥95.0% (by argentometric titration), GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:143-33-9
:
Formula:CNNa
MDL Number:MFCD00003523
ADR:6.1,I
:49.01 g/mol
UN:1689
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Assay (argentometric):min. 95.0 %
- Chloride (Cl):max. 0.05 %
- Hexacyanoferrate:max. 0.05 %
- Sulphide (S):max. 0.02 %
- Lead (Pb):max. 0.005 %
- Identity (IR):Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC87056.290 spec_VWRC87056.360 spec_VWRC87056.460
Product specification
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together