Order Entry
Czech Republic
ContactUsLinkComponent
ɑ-Cyano-4-hydroxycinnamic acid ≥99.0% (by HPLC) matrix substance for MALDI-MS, Supelco®
Irritant
ɑ-Cyano-4-hydroxycinnamic acid ≥99.0% (by HPLC) matrix substance for MALDI-MS, Supelco®
Catalog # 70990-1G-F
Supplier:  Merck
CAS Number:  
Irritant
ɑ-Cyano-4-hydroxycinnamic acid ≥99.0% (by HPLC) matrix substance for MALDI-MS, Supelco®
Catalog # 70990-1G-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance (Color):
    Faint Yellow to Yellow
  • Appearance (Form):
    Powder or Crystals
  • Titration (T) NaOH 0.1M:
    99.0 - 101.0 %
  • Purity (HPLC area %):
    ≥ 99.0 %
  • Melting Point:
    245 - 250 C
  • Solubility (Color):
    Colorless to Faint Green-Yellow
  • Solubility (Turbidity):
    Clear
  • Solubility (Method):
    0.1G IN 10 ml methanol
  • Proton NMR Spectrum:
    Conforms to structure
  • Test by UV Spectroscopy:
    Corresponds to requirements
  • Metal trace Analysis (ICP):
    Corresponds to requirements
  • BARIUM (ICP):
    ≤ 5 mg/kg
  • CALCIUM (ICP):
    ≤ 10 mg/kg
  • CADMIUM (ICP):
    ≤ 5 mg/kg
  • COBALT (ICP):
    ≤ 5 mg/kg
  • CHROMIUM (ICP):
    ≤ 5 mg/kg
  • COPPER (ICP):
    ≤ 5 mg/kg
  • POTASSIUM (ICP):
    ≤ 5 mg/kg
  • MAGNESIUM (ICP):
    ≤ 5 mg/kg
  • MANGANESE (ICP):
    ≤ 5 mg/kg
  • SODIUM (ICP):
    ≤ 10 mg/kg
  • NICKEL (ICP):
    ≤ 5 mg/kg
  • LEAD (ICP):
    ≤ 5 mg/kg
  • ZINC (ICP):
    ≤ 5 mg/kg
  • RECOMMENDED RETEST PERIOD:
    42 MONTHS

Specification Test Results

About this item

 :
 :189.17 g/mol
Melting Pt:245…250 °C (lit.)
 :GHS07
CAS Number:28166-41-8
Formula:C₁₀H₇NO₃
MDL Number:MFCD00004204